Express Eingabe
Austria
ContactUsLinkComponent
Salpetersäure 69% VLSI für Mikroelektronik, J.T.Baker®
Flammable
Corrosive
Irritant
Salpetersäure 69% VLSI für Mikroelektronik, J.T.Baker®
Catalog # 5371-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Salpetersäure 69% VLSI für Mikroelektronik, J.T.Baker®
Catalog # 5371-03
Supplier:  Avantor
CAS Number:  

Specifications

  • VE:
    7 lb
  • Seq:
    7068

Specifications

Specification Test Results

  • For Microelectronic Use:
    69.0 - 70.0 %
  • Assay (HNO₃):
    69.0 - 70.0 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤10
  • Chloride (Cl):
    ≤0.08 ppm
  • Phosphate (PO₄):
    ≤0.05 ppm
  • Sulfate (SO₄):
    ≤0.2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤20 ppb
  • Arsenic and Antimony (as As):
    ≤3.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤50 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤5.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤30.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤5.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤5.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤20.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤30.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤5.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤50.0 ppb
  • Trace Impurities - Potassium (K):
    ≤20.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤60.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤60.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤50.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤90 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤10 par/ml

Specification Test Results

About this item

 :
 :63.01 g/mol
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
UN:2031
ADR:8,II
 :GHS02
 :GHS05
 :GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349