Order Entry
Austria
ContactUsLinkComponent
Nitric acid 65.0 - 68.0%, CMOS for microelectronic, J.T.Baker®
Oxidizing
Corrosive
Irritant
Nitric acid 65.0 - 68.0%, CMOS for microelectronic, J.T.Baker®
Catalog # 9612-33
Supplier:  Avantor
CAS Number:  
Oxidizing
Corrosive
Irritant
Nitric acid 65.0 - 68.0%, CMOS for microelectronic, J.T.Baker®
Catalog # 9612-33
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    7 lb
  • Seq:
    7068

Specifications

Specification Test Results

  • Assay (HNO₃):
    65.0 - 67.9 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤ 10
  • Residue after Ignition:
    ≤ 2 ppm
  • Chloride (Cl):
    ≤ 0.08 ppm
  • Phosphate (PO₄):
    ≤ 0.10 ppm
  • Sulfate (SO₄):
    ≤ 0.2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 40.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 20.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 50 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 30.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 20 ppb
  • Trace Impurities - Gold (Au):
    ≤ 20 ppb
  • Heavy Metals (as Pb):
    ≤ 100 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 40.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 20.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 20 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 20.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 50.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 50 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 50 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 20.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 150.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 30.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 20.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 20.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤ 60 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤ 10 par/ml

Specification Test Results

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
UN:2031
ADR:8,II
 :GHS03
 :GHS05
 :GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349