Order Entry
Austria
ContactUsLinkComponent
n-Heptane, ChromAR® for HPLC, Macron Fine Chemicals™
n-Heptane, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 5410-25
CAS Number:  
undefined
n-Heptane, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 5410-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :100.20 g/mol
UN:1206
ADR:3,II
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544