D-(-)-Mannitol, powder, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- :69-65-8
- :HOH₂C(CH(OH))₄CH₂OH
- :MFCD00064287
- :182.17 g/mol
- :13,05769
- :290…295 °C (4 hPa)
- :164…169 °C
- :1.49 g/cm³ (20 °C)
- :Ambient
- :200-711-8
: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- :69-65-8
- :HOH₂C(CH(OH))₄CH₂OH
- :MFCD00064287
- :182.17 g/mol
- :13,05769
- :290…295 °C (4 hPa)
- :164…169 °C
- :1.49 g/cm³ (20 °C)
- :Ambient
- :200-711-8
Documents
- ACS - Specific Rotation [Å]²⁵ ^D (+):23.3 - 24.3 Degree
- ACS - Insoluble Matter:≤0.01 %
- ACS - Loss on Drying at 105°C:≤0.05 %
- ACS - Residue after Ignition:≤0.01 %
- ACS - Titrable Acid (meq/g):≤0.0008
- ACS - Reducing Sugars:Passes Test
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm



