Order Entry
Austria
ContactUsLinkComponent
 
Magnesium chloride hexahydrate 99.0-105.0%, flakes FCC, Macron Fine Chemicals™
 
undefined
Magnesium chloride hexahydrate 99.0-105.0%, flakes FCC, Macron Fine Chemicals™
 

 

  • Food GMP Manufactured Product:
  • Meets F.C.C. Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Food Chemical:
  • Appearance:
    Passes Test
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Assay (MgCl₂.6H₂O):
    99.0 - 105.0 %
  • FCC - Ammonium (NH₄):
    ≤0.005 %
  • FCC - Lead (Pb):
    ≤4 mg/kg
  • FCC - Sulfate (SO₄):
    ≤0.03 %
  • Solubility:
    Passes Test

 

Frequently Bought Together