Order Entry
Austria
ContactUsLinkComponent
 
 
Lithium chloride ≥99.0% (by argentometric titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
  2370-07
 :  Avantor
 :  
undefined
 
Lithium chloride ≥99.0% (by argentometric titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
  2370-07
 :  Avantor
 :  

 

  • Pack type:
    PE pail, screw top, 3.5 gal
  • Pk:
    12 kg
  • Seq:
    4187

 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (LiCl) (by Ag titrn):
    ≥ 99.0 %
  • ACS - Insoluble Matter:
    ≤ 0.01 %
  • ACS - Loss on Drying:
    ≤ 1.0 %
  • ACS - Titrable Base (meq/g):
    ≤ 0.008
  • ACS - Sulfate (SO₄):
    ≤ 0.01 %
  • ACS - Barium (Ba):
    ≤ 0.002 %
  • ACS - Heavy Metals (as Pb):
    ≤ 0.002 %
  • ACS - Iron (Fe):
    ≤ 0.001 %
  • ACS - Calcium (Ca):
    ≤ 0.01 %
  • ACS - Potassium (K):
    ≤ 0.01 %
  • ACS - Sodium (Na):
    ≤ 0.20 %

 

 

 :
 :42.39 g/mol
 :GHS07
 :Ambient
 :7447-41-8
 :LiCl
 :MFCD00011078