L(+)-Histidine monohydrochloride monohydrate 98.5-101.5% (dried basis), Multi-Compendial, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:5934-29-2
:
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027
:209.63 g/mol
:Management of Change (MOC) category = R
Specification Test Results
- EP/BP - Assay (C₆H₉N₃O₂·HCl) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Identification F:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:3.0 - 5.0
- EP/BP - Specific Rotation [Å]²º^D (+):9.2 - 10.6 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying at 150°C:7.0 - 10.0 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):9.2 - 10.6
- JP - pH:3.5 - 4.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Related Substances:Passes Test
- JP - Water:7.2 - 10.0 %
- JP - Residue on Ignition:≤ 0.10 %
- JP - Assay (anhydrous basis):99.0 - 101.0 %
- Endotoxin Concentration, IU/mg ,For Information Only:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:= 0.028 %
- manufacturing or purification of the product.:= 0.02 %
- Metallic Residues: No metal catalysts or metal reagents, as defined by:= 10 ppm
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:= 10 ppm
- this material.:Passes Test
- Endotoxin Concentration, IU/mg:For Information Only
Specification Test Results
Frequently Bought Together