Order Entry
Austria
ContactUsLinkComponent
Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™
Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # 5189-26
CAS Number:  
undefined
Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # 5189-26
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    200 L
  • Seq:
    5549
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥98.5%
  • For Laboratory,Research,or Manufacturing Use:
    ≤10
  • ACS - Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC):
    ≥98.5 %
  • ACS - Assay (as n-hexane) (by GC), For Information Only:
    ≤0.005%
  • Appearance (clear, colorless liquid):
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Sulfur Compounds (as S):
    ≤0.005 %
  • ACS - Thiophene:
    Passes Test
  • ACS - Water-Soluble Titrable Acid (meq/g):
    ≤0.0003

Specification Test Results

About this item

Synonyms: Hexanes, Isohexane
 :
 :86.18 g/mol
Merck Index:13,04712
Density:0.66 g/cm³ (20 °C)
Flash Pt:–23 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311