Order Entry
Austria
ContactUsLinkComponent
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets J.P. Chemical Specifications:
    Passes Test
  • GMP Manufactured Product:
    ≤0.10%
  • Meets U.S.P Requirements:
    ≤0.10%
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets E.P. Chemical Specifications:
    99.0 - 101.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≥1.249
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B - Diethylene Glycol:
    ≤0.10 %
  • USP - Identification B - Ethylene Glycol:
    ≤0.10 %
  • USP - Identification C:
    Passes Test
  • USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis):
    99.0 - 101.0 %
  • USP - Specific Gravity at 25°/25°C:
    ≥1.249
  • USP - Color:
    Passes Test
  • USP - Residue on Ignition:
    ≤0.01 %
  • USP - Chloride (Cl):
    ≤0.001 %
  • USP - Sulfate (SO₄):
    ≤0.002 %
  • USP - Chlorinated Compounds (as Cl):
    ≤0.003 %
  • USP - Fatty Acids and Esters:
    Passes Test
  • USP - Water (H₂O):
    ≤5.0 %
  • USP - Related Compounds: Individual Impurity:
    ≤0.1 %
  • USP - Related Compo unds: Total Impurities:
    ≤1.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Refractive Index, n²ºD:
    1.470 - 1.475
  • EP/BP - Aldehydes:
    ≤10 ppm
  • EP/BP - Ester:
    Passes Test
  • EP/BP - Halogenated Compounds:
    ≤35 ppm
  • EP/BP - Sugars:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤10 ppm
  • EP/BP - Water (H₂O):
    ≤2.0 %
  • EP/BP - Ash (sulfated):
    ≤0.01 %
  • EP/BP - Impurity A and Related Substances - Impurity A:
    ≤0.1 %
  • EP/BP - Impurity A and Related Substances - Other Impurities with R^T
    ≤0.1 %
  • EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol:
    ≤0.5 %
  • EP - Assay (anhydrous basis):
    98.0 - 101.0 %
  • JP - Assay (anhydrous basis):
    98.0 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Specific Gravity at 20°/20°C:
    ≥1.258
  • JP - Refractive Index, n²ºD:
    ≥1.470
  • JP - Color:
    Passes Test
  • JP - Residue on Ignition:
    ≤0.01 %
  • JP - Chloride (Cl):
    ≤0.001 %
  • JP - Sulfate (SO₄):
    ≤0.002 %
  • JP - Heavy Metals (as Pb):
    ≤5 ppm
  • JP - Fatty Acids and Esters(as butyric acid):
    Passes Test
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Acidity or Alkalinity:
    Passes Test
  • JP - Acrolein, Glucose, and Other Reducing Substances:
    Passes Test
  • JP - Ammonium (NH₄):
    Passes Test
  • JP - Calcium (Ca):
    Passes Test
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Water (H₂O):
    ≤2.0 %
  • JP - Ethylene Glycol:
    ≤0.1 %
  • JP - Diethylene Glycol:
    ≤0.1 %
  • JP - Any Other Impurity:
    ≤0.1 %
  • JP - Total Impurities:
    ≤1.0 %
  • Endotoxin Concentration (EU/g):
    ≤5

Specification Test Results

Frequently Bought Together