Order Entry
Austria
ContactUsLinkComponent
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, J.T.Baker®
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    99.0 - 101.0%
  • Meets U.S.P Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.10 %
  • Bulk Pharmaceutical Chemical:
    ≤0.10 %
  • USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis):
    99.0 - 101.0 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B - Diethylene Glycol:
    ≤0.10 %
  • USP - Identification B - Ethylene Glycol:
    ≤0.10 %
  • USP - Identification C:
    Passes Test
  • USP - Specific Gravity at 25°/25°C:
    ≥1.249
  • USP - Color:
    Passes Test
  • USP - Residue on Ignition:
    ≤0.01 %
  • USP - Chloride (Cl):
    ≤0.001 %
  • USP - Sulfate (SO₄):
    ≤0.002 %
  • USP - Chlorinated Compounds (as Cl):
    ≤0.003 %
  • USP - Fatty Acids and Esters:
    Passes Test
  • USP - Water (H₂O):
    ≤5.0 %
  • USP - Related Compounds: Individual Impurity:
    ≤0.1 %
  • USP - Related Compo unds: Total Impurities:
    ≤1.0 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together