Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Formalin solution 10%, BAKER, neutralised, J.T.Baker®

20 L
Catalog # 7385.9020
Supplier:   Avantor
Formalin solution 10%, BAKER, neutralised, J.T.Baker®
Catalog # 7385.9020
Supplier:  Avantor
CAS Number:  50-00-0
Formalin solution 10%, BAKER, neutralised, J.T.Baker®

20 L

CAS Number:  50-00-0
Price
97,27 €
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • UN:2209
  • ADR:8,III

 

Specifications

Specification Test Results

  • Formaldehyde (HCHO):
    38 - 42 g/l
  • pH at 25°C:
    6.8 - 7.2

Specifications

  • Pk:
    20 L

Specifications