Order Entry
Austria
ContactUsLinkComponent
Ethyl acetate, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Ethyl acetate, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
  5277.2500
 :  Avantor
   Safer Human and Environmental Health
 :  
undefined
Ethyl acetate, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
  5277.2500
 :  Avantor
   Safer Human and Environmental Health
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay (by GC):
    min. 99.8 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Water (KF):
    max. 0.05 %

 

 

 :
 :88.11 g/mol
 :1173
 :3,II
 :141-78-6
 :CH₃COOC₂H₅
 :MFCD00009171