Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7125.1000
Supplier:  Avantor
CAS Number:  
EDTA disodium salt, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7125.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.0195 - 0.0205

Specification Test Results

About this item

 :
 :336.21 g/mol
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672