Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt dihydrate 99.01-101.0% (dried basis), crystals USP, J.T.Baker®
EDTA disodium salt dihydrate 99.01-101.0% (dried basis), crystals USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
EDTA disodium salt dihydrate 99.01-101.0% (dried basis), crystals USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    4.0 - 6.0
  • USP - Loss on Drying:
    8.7 - 11.4 %
  • USP - Calcium (Ca):
    Passes Test
  • USP - Nitrilotriacetic Acid:
    ≤0.1 %
  • USP - Assay (C₁₀H₁₄N₂Na₂O₈) (dried basis):
    99.0 - 101.0 %

Specification Test Results

Related Information

Frequently Bought Together