Order Entry
Austria
ContactUsLinkComponent
Diethyl ether ≥99.5% (by GC, corrected for water content) stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Diethyl ether ≥99.5% (by GC, corrected for water content) stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2854-25
CAS Number:  
undefined
Diethyl ether ≥99.5% (by GC, corrected for water content) stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2854-25
CAS Number:  

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay ((C₂H₅)₂O) (by GC, exclusive of preservative, corrected for water):
    ≥99.5 %
  • Alcohol (C₂H₅OH)(by GC) (preservative)(v/v):
    1.5 - 2.5 %
  • Color (APHA):
    ≤10
  • Peroxide (as H₂O₂)(at time of packaging):
    ≤0.0005 %
  • Optical Absorbance (1-cm path vs water) - 215 nm:
    ≤1.00
  • Optical Absorbance (1-cm path vs water) - 230 nm:
    ≤0.30
  • Optical Absorbance (1-cm path vs water) - 254 nm:
    ≤0.08
  • Optical Absorbance (1-cm path vs water) - 280 nm:
    ≤0.01
  • Optical Absorbance (1-cm path vs water) - 300 nm:
    ≤0.005
  • Residue after Evaporation:
    ≤0.0005 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %

Specification Test Results

About this item

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
 :Stabilised with ethanol 1.5 - 2.5%
REACH:01-2119535785-29
 :GHS02
 :GHS07
 :GHS08
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646