

Dichloromethane, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Boiling Pt:39.8 °C (1013 hPa)
UN:1593
ADR:6.1,III
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together