Order Entry
Austria
ContactUsLinkComponent
 
Irritant
Health
Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # 4881-26
CAS Number:  
undefined
Irritant
Health
Dichloromethane ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # 4881-26
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    ≥99.5%
  • ACS - Assay (CH₂Cl₂) (by GC):
    ≥99.5 %
  • Appearance (clear, colorless liquid):
    Passes Test
  • ACS - Carbon Tetrachloride (mg/L):
    ≤10.0000 ppm
  • ACS - Color (APHA):
    ≤10
  • ACS - Free Halogens:
    Passes Test
  • ACS - Residue after Evaporation:
    ≤0.002 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Water (H₂O)(by coulometry):
    ≤0.02 %

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881