Order Entry
Austria
ContactUsLinkComponent
Copper(II) chloride dihydrate ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™
Corrosive
Environment
Copper(II) chloride dihydrate ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 4824-20
CAS Number:  
undefined
Corrosive
Environment
Copper(II) chloride dihydrate ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 4824-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (CuCl₂·2H₂O) (by iodometry):
    ≥99.0 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.005 %
  • ACS - Nitrate (NO₃):
    ≤0.015 %
  • Solubility:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Sodium (Na):
    ≤0.02 %
  • ACS - Nickel (Ni):
    ≤0.01 %
  • ACS - Potassium (K):
    ≤0.01 %
  • ACS - Calcium (Ca):
    ≤0.005 %

Specification Test Results

About this item

 :
 :170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
 :GHS05
 :GHS09
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674