Chloroform, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
UN:1888
ADR:6.1,III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together