Order Entry
Austria
ContactUsLinkComponent
Orthoboric acid ≥99.5%, granules, GenAR® NF, Macron Fine Chemicals™
Irritant
Health
Orthoboric acid ≥99.5%, granules, GenAR® NF, Macron Fine Chemicals™
Catalog # 7779-06
CAS Number:  
undefined
Irritant
Health
Orthoboric acid ≥99.5%, granules, GenAR® NF, Macron Fine Chemicals™
Catalog # 7779-06
CAS Number:  

Specifications

  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    99.5 - 100.5 %
  • NF - Assay (H₃BO₃) (dried basis):
    99.5 - 100.5 %
  • NF - Completeness of Solution:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Loss on Drying:
    ≤0.5 %
  • NF - Solubility in Alcohol:
    Passes Test
  • NF - LAL Bacterial Endotoxins (EU/g):
    ≤35
  • EP/BP - Assay (H₃BO₃):
    99.0 - 100.5 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - pH of a 3.3% Solution:
    3.8 - 4.8
  • EP/BP- Organic Matter:
    Passes Test
  • EP/BP - Solubility in Alcohol:
    Passes Test
  • EP/BP - Sulfate (SO₄):
    ≤450 ppm
  • JP - Assay (H₃BO₃) (dried basis):
    99.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Arsenic (As):
    ≤5 ppm
  • JP - Loss on Drying:
    ≤0.5 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    = 0.5 %

Specification Test Results

About this item

 :
Boiling Pt:185 °C (1013 hPa)
 :Management of Change (MOC) category = R
 :61.83 g/mol
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337