Order Entry
Austria
ContactUsLinkComponent
Orthoboric acid 99.5-100.5% (dry basis), granular NF, Multi-Compendial, J.T.Baker®
Irritant
Health
Orthoboric acid 99.5-100.5% (dry basis), granular NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Health
Orthoboric acid 99.5-100.5% (dry basis), granular NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • NF - Completeness of Solution:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Solubility in Alcohol:
    Passes Test
  • NF - Loss on Drying:
    ≤ 0.5 %
  • NF - Assay (H₃BO₃) (dried basis):
    99.5 - 100.5 %
  • NF - Endotoxin Concentration (EU/g):
    ≤ 35
  • EP/BP - Assay (H₃BO₃):
    99.0 - 100.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    3.8 - 4.8
  • EP/BP - Solubility in Alcohol:
    Passes Test
  • EP/BP - Organic Matter:
    Passes Test
  • EP/BP - Sulfate (SO₄):
    ≤ 450 ppm
  • JP - Assay (H₃BO₃) (dried basis):
    99.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Arsenic (As):
    ≤ 5 ppm
  • JP - Loss on Drying:
    ≤ 0.5 %

Specification Test Results

Related Information

Frequently Bought Together