Order Entry
Austria
ContactUsLinkComponent
Ammonium sulphate 99.0-100.5%, granular NF, ACS, J.T.Baker®
 
Ammonium sulphate 99.0-100.5%, granular NF, ACS, J.T.Baker®
  3523-65
 :  Avantor
 :  
undefined
 
Ammonium sulphate 99.0-100.5%, granular NF, ACS, J.T.Baker®
  3523-65
 :  Avantor
 :  

 

  • Pack type:
    HDPE drum, Flowmor™ package
  • Pk:
    100 kg
  • Seq:
    4553

 

 

  • Bulk Pharmaceutical Chemical:
    99.0 - 100.5 %
  • NF - Assay ((NH₄)₂SO₄):
    99.0 - 100.5 %
  • NF - Insoluble Matter:
    ≤0.005 %
  • NF - Residue on Ignition:
    ≤0.005 %
  • NF - pH of 5% Solution:
    5.0 - 6.0
  • NF - Nitrate (NO₃):
    ≤0.001 %
  • NF - Chloride (Cl):
    ≤5 ppm
  • NF - Phosphate (PO₄):
    ≤5 ppm
  • NF - Iron (Fe):
    ≤5 ppm
  • NF - Identification:
    Passes Test
  • NF - Total Aerobic Microbial Count:
    ≤100 cfu/g
  • NF - Total Yeast and Mold Count:
    ≤10 cfu/g
  • ACS - Assay ((NH₄)₂SO₄) (A.C.S.):
    ≥99.0 %
  • ACS - Residue on Ignition:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 6.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤3 ppm
  • ACS - Nitrate (NO₃):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤3 ppm
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • Appearance (White crystals or granules):
    Passes Test

 

 

 :
 :132.14 g/mol
 :1.77 g/cm³
 :GHS08
 :7783-20-2
 :(NH₄)₂SO₄
 :MFCD00003391