Ammonia solution 29%, Finyte® for microelectronic, J.T.Baker®
Artikelnr. 5817-09
Supplier: Avantor
CAS Number:
Specifications
- Pk:400 lb
- Seq:7284
Specifications
Specification Test Results
- Appearance:Passes Test
- Assay (as NH₃):28.0 - 30.0 %
- Color (APHA):≤10
- Residue after Ignition:≤3 ppm
- Carbon Dioxide (CO₂):≤10 ppm
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.2 ppm
- Nitrate (NO₃):≤0.4 ppm
- Phosphate (PO₄):≤0.10 ppm
- Total Sulfur (as SO₄):≤0.2 ppm
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Arsenic and Antimony (as As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤5.0 ppb
- Trace Impurities - Calcium (Ca):≤20 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤1.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Silicon (Si):≤10.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤10.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count at point of fill - 0.5 µm and greater:≤150 par/ml
- Particle Count at point of fill - 1.0 µm and greater:≤25 par/ml
Specification Test Results
About this item
:
:35.05 g/mol
UN:2672
ADR:8,III
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650