Order Entry
Austria
ContactUsLinkComponent
 
Acetonitrile ≥99.9%, ChromAR® HPLC super gradient, Macron Fine Chemicals™
Catalog # 2856-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Acetonitrile ≥99.9%, ChromAR® HPLC super gradient, Macron Fine Chemicals™
Catalog # 2856-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (CH₃CN):
    ≥ 99,9 %
  • Water (KF):
    ≤ 0,02 %
  • Titrable Acid (meq/g):
    ≤ 0,0005
  • Titrable Base (meq/g):
    ≤ 0,0002
  • Residue after Evaporation:
    ≤ 0,0005%
  • 195 mm:
    ≤ 0,12
  • 200 nm:
    ≤ 0,03
  • 210 nm:
    ≤ 0,02
  • 220 nm:
    ≤ 0,01
  • 254 - 400 nm :
    ≤ 0,005
  • Gradient Elution Test 210 nm:
    ≤ 3 mAU

Specification Test Results

About this item

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
 :
 :41.05 g/mol
Melting Pt:–45.7 °C
UN:1648
REACH:01-2119471307-38
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6 °C (1013 hPa)
Density:0.782 g/cm³ (20 °C)
Flash Pt:2
ADR:3 II
Merck Index:13,00071