Order Entry
Austria
ContactUsLinkComponent
Acetone, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Acetone, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5276.2500
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Acetone, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5276.2500
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.8 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Water (KF):
    max. 0.3 %

Specification Test Results

About this item

 :
 :58.08 g/mol
UN:1090
ADR:3,II
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765