Express Eingabe
Austria
ContactUsLinkComponent
 
2-Propanol, Finyte® für Mikroelektronik, J.T.Baker®
  5873-15
 :  Avantor
 :  
undefined
2-Propanol, Finyte® für Mikroelektronik, J.T.Baker®
  5873-15
 :  Avantor
 :  

 

  • VE:
    4 L
  • Seq:
    5162

 

 

  • For Microelectronic Use:
    = 99.9 %
  • Assay (CH₃CHOHCH₃):
    ≥ 99.9 %
  • Color (APHA):
    ≤ 6
  • Residue after Evaporation:
    ≤ 3000.0 ppb
  • Solubility in H₂O:
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤ 0.05 %
  • Acidity (µeq/g):
    ≤ 0.2
  • Alkalinity (µeq/g):
    ≤ 0.1
  • Chloride (Cl):
    ≤ 0.2 ppm
  • Phosphate (PO₄):
    ≤ 75.0 ppb
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 10.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 3.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 5.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 5.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 10.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 5.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤ 25 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤ 8 par/ml

 

 

 :
 :60.1 g/mol
 :3,II
 :Veränderungsmanagement-Kategorie= R
 :1219
 :67-63-0
 :(CH₃)₂CHOH
 :MFCD00011674