1,4-Dioxane, AR®, Laboratory Reagent, Macron Fine Chemicals™
Catalog # 5413-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
Specifications
About this item
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
:
:88.11 g/mol
UN:1165
ADR:3,II
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571