Membrane disc filters, Versapor® Acrylic Copolymer
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Hydrophilic acrylic polymer Versapor® membrane on non woven support ideal for pre-filtration of difficult to filter solutions and serum.
- Reduces clogging of final filters. Can be used upstream of a sterilising filter for viscous solutions
- Strong and flexible while both wet or dry
- Excellent throughput with high liquid flow rates at low differential pressures
Designed to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar.
UV or gamma sterilisable.
Specifications
- Catalog No:
- 514-4207
- 516-9020
- 514-1220
- 516-9018
- 514-4208
- 514-4210
- 514-4209
- 514-0749
- 516-7890
- 514-4214
- 514-4203
- 514-4204
- 514-4205
- 514-4215
- 514-4206
- Type:
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Versapor®
- Thickness:
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- 94 µm
- Water flow rate:
- 315 ml/min/cm² (0,7 bar)
- 75 ml/min/cm² (0,7 bar)
- -
- 315 ml/min/cm² (0,7 bar)
- 315 ml/min/cm² (0,7 bar)
- 142 ml/min/cm² (0,7 bar)
- 142 ml/min/cm² (0,7 bar)
- 778 ml/min/cm² (0,7 bar)
- 778 ml/min/cm² (0,7 bar)
- 75 ml/min/cm² (0,7 bar)
- 518 ml/min/cm² (0,7 bar)
- 518 ml/min/cm² (0,7 bar)
- 518 ml/min/cm² (0,7 bar)
- 75 ml/min/cm² (0,7 bar)
- 315 ml/min/cm² (0,7 bar)
- Max. temperature:
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
- 88 °C
Specifications
Frequently Bought Together