Orders LinkContactUsLinkComponent
Antioxidant assay kit
Antioxidant assay kit
Antioxidant Assay Kit, 100 assays, 1 KIT
Catalog # ABNOKA1622
Supplier:   Abnova
Antioxidant assay kit
Catalog # ABNOKA1622
Supplier:  Abnova
CAS Number:  
Antioxidant assay kit
Antioxidant assay kit
Antioxidant Assay Kit, 100 assays, 1 KIT
CAS Number:  
Price
648,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Antioxidant Assay Kit is a quantitative colorimetric determination of total antioxidant capacity of cellular samples.
  • Sensitive and accurate, linear detection range from 1,5 to 1000 µM Trolox equivalents
  • Simple procedure involves addition of a single working reagent and incubation for 10 min
  • Can be readily automated as a high-throughput assay for thousands of samples per day
Antioxidant Assay Kit measures total antioxidant capacity in which Cu²⁺ is reduced by antioxidant to Cu⁺. The resulting Cu⁺ specifically forms a colored complex with a dye reagent. The color intensity at 570 nm is proportional to TAC in the sample.
Antioxidant assay kit is suitable for serum, plasma, urine, saliva and other biological samples, food and beverages. Also used in drug discovery/pharmacology to check the effect of drugs on TAC.
Delivery information: Kit includes Reagent A, 12 ml; Reagent B, 1 ml; and Standard (50 mM Trolox, 100 µl).
Documents
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Antioxidant Assay Kit, 100 assays