Standard hydrometers, M100
About this item
These specific gravity hydrometers are marked with reference norms, calibration temperature, surface tension, individual and serial numbers.
- Reading below the meniscus
- Calibration temperature of 15 °C
- Benchmark on the stem to show any movement of the scale
Calibrated according to the following tensions:
0,600 to 1,000 Sp.Gr.= 16 to 35 mN/m (low tension)
1,000 to 1,300 Sp.Gr.= 55 mN/m (medium tension)
: ISO 650.
: 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate is available on request. Includes a Traceability and Conformity Certificate.
11 Options Available Below
Product Details & Documents
These specific gravity hydrometers are marked with reference norms, calibration temperature, surface tension, individual and serial numbers.
- Reading below the meniscus
- Calibration temperature of 15 °C
- Benchmark on the stem to show any movement of the scale
Calibrated according to the following tensions:
0,600 to 1,000 Sp.Gr.= 16 to 35 mN/m (low tension)
1,000 to 1,300 Sp.Gr.= 55 mN/m (medium tension)
: ISO 650.
: 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate is available on request. Includes a Traceability and Conformity Certificate.
Product Family Options
Product Information
- ModelRangeDivisionLengthAvailabilityPrice