Anti-VSV-G tag Mouse Monoclonal Antibody [clone: 8D6]
ABNOMAB15245
: Abnova
- Antibody type:Primary
- Antigen symbol:VSV-G tag
- Clonality:Monoclonal
- Clone:8D6
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Storage buffer:In PBS (50% glycerol, 0.5% BSA, 0.02% sodium azide).
- Storage temperature:Store at -20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to full length of human VSV-G tag.
- Purification:Affinity purification
- Size:100 µL
- Pk:100 µl
Mouse monoclonal antibody raised against synthetic peptide of human VSV-G tag.
Type: Primary
Antigen: VSV-G tag
Clonality: Monoclonal
Clone: 8D6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: