Hemoglobin reagent
Catalog # DIAS225499910021
Supplier: DIAGNOSTIC SYSTEMS DIASYS
CAS Number:
Specifications
- Pk:1 KIT
- Description:Hemoglobin reagent
- Environmentally Preferable:
- Volume:80 ml (2 x 20 mL + 2 x 20 mL)
Specifications
About this item
Reagent for in vitro diagnostics (IVD) in human samples.
- Liquid in form
- Fluid-stable
Suitable for use in clinical chemistry.