Equilibrium DIALYZER™ plate for Single Use 96-Well Equilibrium DIALYZER™
Catalog # HARV74-2331
Supplier: HARVARD APPARATUS
Specifications
- Description:96-Well Equilibrium DIALYZER™ plate with 10 KDa MWCO membrane
- Pk:1
Specifications
About this item
The Single Use 96-Well Equilibrium DIALYZER™ is designed for high throughput protein interaction studies.
- Each well in this system has a separate membrane, eliminating possibility of cross-contamination
- Excellent sample recovery
- Wells are sealed with 8-cap strips (not included)