ContactUsLinkComponent
VWR®, Cap Mat Applicator
VWR®, Cap Mat Applicator
Catalog # 732-3335
Supplier:  VWR Collection
VWR®, Cap Mat Applicator
Catalog # 732-3335
Supplier:  VWR Collection
Specifications
  • Description:
    Cap mat applicator
  • Seq:
    05
  • Pk:
    1
Specifications
About this item
Compact and portable, the cap mat applicator is very easy to use, requiring only one operation of the system to produce an accurate and tight seal on a wide range of both collection and shallow well microplates.
  • Needs minimal pressure for capping, reducing fatigue and RSI
  • Powder coated to resist chemical spillage