VWR®, Deep Well Plates, 96 Round Well
Supplier: VWR Collection
Certificates
About this item
Made of virgin PP to minimise extractables.
- Raised rim to prevent cross contamination and enable a better heat-welded seal
- Also available rimless to allow insertion of vials, or when adhesive seals are used
- Free from DNase and RNase
- Round bottom for optimal mixing and recovery
- Small volumes with flat, U- or V-bottoms are also available
- Can be stored at –80 °C
Suitable for use with automated sample handling systems.
Frequently Bought Together