Order Entry
Austria
Orders LinkContactUsLinkComponent
Electrode storage solution

Electrode storage solution

pH/ORP electrode storage solution, 500 ml bottle, 500 mL
Catalog # HANNHI70300L
Supplier:   Hanna
Electrode storage solution
Catalog # HANNHI70300L
Supplier:  Hanna
CAS Number:  
Electrode storage solution

pH/ORP electrode storage solution, 500 ml bottle, 500 mL

CAS Number:  
Price
25,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

HI 70300L is a storage solution prepared with reagent grade chemicals which can be used to ensure maximum performance of pH and ORP electrodes.

  • Keeps the glass membrane of the pH electrode hydrated
  • Manufactured specifically for pH and ORP electrodes
  • Air tight bottle with tamper-proof seal ensures quality and freshness of solution
  • Clearly marked expiration date and lot number

The storage solution is specifically formulated to minimise microbial growth and to prevent any effects of diffusion/osmotic from storing a probe in a solution with the highly concentrated inner reference electrolyte.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    pH/ORP electrode storage solution, 500 ml bottle

Specifications