Order Entry
Austria
ContactUsLinkComponent
Ammonia solution ≥25% in water eluent additive, for LC-MS, Fluka™
Irritant
Corrosive
Ammonia solution ≥25% in water eluent additive, for LC-MS, Fluka™
Certificates
Irritant
Corrosive
Ammonia solution ≥25% in water eluent additive, for LC-MS, Fluka™

Specification Test Results

  • Assay:
    Min. 25 %
  • Density (D 20/4):
    0.890 - 0.910
  • Non-volatile matter:
    Max. 0.001 %
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.01 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.01 ppm
  • Calcium (Ca):
    Max. 0.01 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Cesium (Cs):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.01 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Germanium (Ge):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.01 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.01 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.01 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.01 ppm
  • Rubidium (Rb):
    Max. 0.01 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.01 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.01 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 0.2 ppm
  • Phosphate (PO₄):
    Max. 0.2 ppm
  • Sulfate (SO₄):
    Max. 1 ppm
  • Suitability for the UHPLC-MS:
    complying
  • Storage temperature:
    2 - 8 °C

Specification Test Results

Related Information

Frequently Bought Together