Plastic ACME threaded handles
About this item
Lightweight, plastic coated fiberglass handles with threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
- Lightweight, plastic coated fiberglass handles
- ACME Connect threaded end connection
- Hand grips available in four color options for colour coding
- Available fixed length or extendable
- Fixed lengths range from 300 mm to 1,5 m
- Extendable maximum lengths range from 900 mm to 3,5 m
- ACME Connect threaded end attaches directly to plastic threaded mops, tools and adapters
- SSA-2 adapter connects threaded handles to Quick Disconnect mops, tools and adapters
- Good compatibility with disinfectants
- Autoclavable
The Micronova Plastic ACME Threaded Handles attach directly to the plastic (QP series) adapters and tools. Use the SSA-2 ACME Thread Adapter to attach to stainless steel Quick Disconnect (QD series) adapters and tools.
9 Options Available Below
Product Details & Documents
Lightweight, plastic coated fiberglass handles with threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
- Lightweight, plastic coated fiberglass handles
- ACME Connect threaded end connection
- Hand grips available in four color options for colour coding
- Available fixed length or extendable
- Fixed lengths range from 300 mm to 1,5 m
- Extendable maximum lengths range from 900 mm to 3,5 m
- ACME Connect threaded end attaches directly to plastic threaded mops, tools and adapters
- SSA-2 adapter connects threaded handles to Quick Disconnect mops, tools and adapters
- Good compatibility with disinfectants
- Autoclavable
The Micronova Plastic ACME Threaded Handles attach directly to the plastic (QP series) adapters and tools. Use the SSA-2 ACME Thread Adapter to attach to stainless steel Quick Disconnect (QD series) adapters and tools.
Documents
Product Family Options
Extendable handles
Product Information
- LengthAvailabilityPrice
- MICRSTAEH-036WHPlastic ACME threaded extendable handle, with white hand gripPlastic ACME threaded extendable handle, with white hand grip460 - 910 mmLog in for availability1 * 1 ST164,00 €
- MICRSTAEH-072WHPlastic ACME threaded extendable handle, with white hand gripPlastic ACME threaded extendable handle, with white hand grip102 - 183 cmLog in for availability1 * 1 ST164,00 €
- MICRSTAEH-101WHPlastic ACME threaded extendable handle, with white hand gripPlastic ACME threaded extendable handle, with white hand grip1016 - 2565 mmLog in for availability1 * 1 ST255,00 €
- 129-1075Plastic ACME threaded extendable handle, with white hand gripPlastic ACME threaded extendable handle, with white hand grip1727 - 4650 mmLog in for availability1 * 1 ST321,00 €
- MICRSTAEH-237BLPlastic ACME threaded extendable handle, with black hand gripPlastic ACME threaded extendable handle, with black hand grip1727 - 6020 mmLog in for availability1 * 1 ST406,00 €
Fixed length handles
Product Information
- LengthAvailabilityPrice
- MICRSTAH-24WHPlastic ACME threaded handle, with white hand grip
- MICRSTAH-36WHPlastic ACME threaded handle, with white hand grip
- MICRSTAH-48WHPlastic ACME threaded handle, with white hand grip
- MICRSTAH-60WHPlastic ACME threaded handle, with white hand grip
Fixed length handles
Product Information
- Catalog #DescriptionLengthAvailabilityPrice
- Plastic ACME threaded extendable handle, with white hand grip305 mmLog in for availability1 * 1 ST30,50 €
Adapter
Product Information
- Catalog #DescriptionAvailabilityPrice