ContactUsLinkComponent
Wipes, KIMTECH PREP* WETTASK*
Wipes, KIMTECH PREP* WETTASK*
Catalog # 115-1997
Supplier:  KIMBERLY CLARK
undefined
Wipes, KIMTECH PREP* WETTASK*
Catalog # 115-1997
Supplier:  KIMBERLY CLARK
Specifications
  • Description:
    DSX wipes, 90 sheets/roll
  • L×W:
    318×305 mm
  • Seq:
    03
  • Pk:
    6 Roll
Specifications
About this item
This refillable wet wiping system is the ideal alternative to using a spray bottle or open bucket in combination with a rag, making it easy to use and versatile. Two different types, depending on application.
  • Closed-bucket system keeps the pre-moistened wipes fresh in the container and avoids contamination
  • Designed to be compatible with quaternary amine disinfectants
  • Reduces exposure to chemical vapours, splashes and spills
SXX for working with solvents, very strong and absorbent rayon/polyester, no loose fibres. Solvent resistant won't break down when wet. Low lint level suitable for critical tasks.
DS/DSX for use with sanitisers/disinfectants. Leaves some disinfectant/sanitiser on the surface that has been wiped allowing it to work throughly.
Colour: White.