Order Entry
Austria
Orders LinkContactUsLinkComponent
Sludge nabber

Sludge nabber

Sludge nabber, up to 200 cm
Catalog # NASCB01229WA
Supplier:   NASCO FORT ATKINSON
Sludge nabber
Catalog # NASCB01229WA
Sludge nabber
Sludge nabber

Sludge nabber, up to 200 cm

Price
339,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sample bottles of various sizes are held via an adjustable locking device.

  • Makes sample collection of liquids fast and easy
  • Flexible with adjustable aluminum pole
  • PU holder with a plastic snapper ring

Suitable for numerous requirements including chlorine in solution – Can also be used for BOD.

Delivery information: Supplied with a 500 ml PE bottle.

Documents

Specifications
  • Description:
    Sludge nabber, up to 200 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.