Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nucleic Acid Reagents
Nucleic Acid Modification and Labelling
Hybrisol VII
Hybrisol VII
Hybrisol VII
Catalog # ICNA11RIST139010
Supplier:
MP Biomedicals
CAS Number:
Expand image
Hybrisol VII
Catalog # ICNA11RIST139010
Supplier:
MP Biomedicals
CAS Number:
About this item
Product Details
Product Details