Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator, for COD determination (low range)
Catalog # HACH2055133
Supplier: Hach
CAS Number:
Specifications
- Pack type:Dropping bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:692.54 g/mol
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428