Order Entry
Austria
ContactUsLinkComponent
Cleaning solution, DNA-OFF™, MP Biomedicals
Cleaning solution, DNA-OFF™, MP Biomedicals
Catalog # ICNA11QD0500
Supplier:  MP Biomedicals
CAS Number:  
Cleaning solution, DNA-OFF™, MP Biomedicals
Catalog # ICNA11QD0500
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    DNA-OFF™ Non alkaline cleaning solution
  • pH value:
    8 - 9

Specifications

About this item

DNA-OFF™ solution contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors.

  • Used for eliminating DNA from all surfaces
  • It is stable and heat resistant

DNA-OFF™ is a ready to use non-corrosive solution and does not contain carcinogenic compounds.