Safety spectacles, 2840 series
Lieferant: 3M
Zertifikate
Über diesen Artikel
These single lens safety glasses are available in different lens options: clear, yellow for improved contrast in low light conditions, smoke for protection against sun glare, light gold mirror for indoor/outdoor work, infra red shade five for certain gas welding/cutting and brazing applications and red-orange for protection against intense blue light.
- Optical class 1 PC lens for highest optical clarity and extended wear time
- Adjustable lens angle for individual fit
- Adjustable temple length (4 positions) for wearer comfort and optimal fit
- Integrated brow guard for increased protection
- Soft inner padding on the temples for increased stability
Weight: 26 g
Certifications: EN 166, EN 170/172
Zugehörige Informationen
Frequently Bought Together