Disposable particle filtering respirators, FFP3, healthcare, Aura™ 1883+
Artikelnr. 111-1525
Lieferant: 3M
Spezifikationen
- Type:1883+
- Size:U*
- Environmentally Preferable:
- Pk:8
- With valve:+
- Class:FFP3
Spezifikationen
Über diesen Artikel
Unique FFP3 respirator with two-way protection and added exhalation valve for comfort.
- Three panel design for increased comfort with the added convenience of an individually wrapped foldable product
- Includes shrouded valve fitted to the front of the respirator, which prevents heat and moisture build-up from exhaled breath
- Meets requirements and testing for Bacterial Filtering Efficiency (BFE) and splash resistance
- Red colour coded straps to denote FFP3 protection level
Certifications: Highest Protection Level. Assigned Protection Factor = 20
Caution: *One size fits all