Order Entry
Austria
Orders LinkContactUsLinkComponent
Eyewash solution, pH neutral

Eyewash solution, pH neutral

Eyewash solution, pH neutral
Supplier:  Plum Safety
Certificates
Eyewash solution, pH neutral
Eyewash solution, pH neutral

2 Options Available Below

Product Details & Documents

Sterile phosphate buffer solution (4,9%) for fast and effective neutralisation of acids and alkalis.

  • Handy sized bottle, well suited for use in first aid kits and emergency bags
  • Bottle with ergonomic eye cup, dust cap and clear instructions
  • Can be used as refill for Plum wall-mounted eyewash station
  • No maintenance and cleaning of the bottles required
  • Bottles in single carton: Carton protects the bottles from external contaminations and can be opened quickly and easily at the perforation

Rinsing time: ~2 minutes per 200 ml
Shelf life: 3 years

According to DIN EN 15154-4

Certifications: According to DIN EN 15154-4.

Product Family Options

Product Information

  • Availability
    Price
  • 118-1707
    Eyewash solution, pH neutral, 200 ml bottle
    Eyewash solution, pH neutral, 200 ml bottle
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,20 €
    15,20 €
    1 * 1 ST
       15,20 €

  • 118-0479
    Eyewash solution, pH neutral, 200 ml, bottle in single carton
    Eyewash solution, pH neutral, 200 ml, bottle in single carton
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,70 €
    15,70 €
    1 * 1 ST
       15,70 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Belt bag for 200 ml eyewash bottles
    Belt bag for 200 ml eyewash bottles
    118-1704
    Log in for availability
    1 * 1 ST - 8,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    118-1712
    Log in for availability
    1 * 1 ST - 4,50 €

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.