ContactUsLinkComponent
Adhesive seal, VIEWseal™
Adhesive seal, VIEWseal™
Catalog # 736-0232
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
undefined
Adhesive seal, VIEWseal™
Catalog # 736-0232
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Optical property:
    Transparent
  • Description:
    VIEWseal™
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    100
Specifications
About this item
VIEWseal™ is an adhesive sealing film coated with a silicone adhesive, which only sticks when the film surface is pressed on. The film is easy to work with, even when wearing gloves (the film does not adhere to gloves).
  • Temperature resistant from −80 to 110 °C
  • Highly transparent for precise optical measurements
  • Ideally suited for protein crystallography