Order Entry
Austria
ContactUsLinkComponent
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Catalog # ICNA11RIST2050
Supplier:  MP Biomedicals
CAS Number:  
Buffer, SSC 20X powder for molecular biology, MP Biomedicals
Catalog # ICNA11RIST2050
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    SSC buffer solid for 20X concentrated solution

Specifications

About this item

SSC buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates. SSC is used as a hybridisation buffer in immunoblotting and DNA microarray protocols.