Protein isolation and purification, FastPROTEIN™ blue matrix kit, MP Biomedicals
Artikelnr. ICNA116550400
Lieferant: MP Biomedicals
CAS-Nummer:
Spezifikationen
- Pk:50
- Description:FastPROTEIN™ Blue Matrix Kit
Spezifikationen
Über diesen Artikel
The FastPROTEIN™ Blue Matrix Kit (also known as Lysing Matrix B) is designed for rapid isolation and purification of protein from bacterial cultures.
- Easy lysis of bacterial cells to release recombinant protein
- Used with FastPrep® instruments to reproducibly lyse gram positive and gram negative bacterial cells within 40 seconds
Rapid release of recombinant protein saves valuable time during optimisation of expression. Information from up to 24 different induction variables can be quickly obtained after one 40 second FastPrep® run followed by electrophoresis. Once the protein of interest is expressed and identified, the active proteins released by the unique motion of the FastPrep® instruments can be recovered from the lysate by a wide range of protein-specific techniques.
Delivery: Kit contains matrix B, 0,1 mm silica (glass) and 50 tubes.