Ballotini solid soda glass balls, SiLibeads®
Supplier: SIGMUND LINDNER
Certificates
About this item
Ballotini solid glass balls are produced from high quality and pure soda-lime glass.
- Main applications include:
- Chemical plants (where large inert areas are required) and filters, example fractionating columns
- Resin filler
- Shot blasting
- Burnishing (metal finishing)
- Gas chromatography
The unique washing and polishing process does not involve the addition of hydrofluoric acid and thus gives the balls a pure shiny surface without contamination.
Frequently Bought Together